Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD226 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587772 Supplier Invitrogen™ Supplier No. PA587772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEVLWHTSVP FAENMSLECV YPSMGILTQV EWFKIGTQQD SIAIFSPTHG MVIRKPYAER VYFLNSTMAS NNMTLFFRNA SEDDVGYYSC SLYTYPQGTW QKVIQVVQSD SFEAAVPSNS HIVSEPGKNV TLTCQPQMTW PVQAVRWEKI QPRQIDLLTY CNLVHGRNFT SKFPRQIVSN CSHGRWSVIV IPDVTVSDSG LYRCYLQASA GENETFVMRL TVAEGKTDNQ YTLFVA; Positive Samples: Mouse lung; Cellular Location: Membrane, Single-pass type I membrane protein.

DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD226
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cd226
Gene Accession No. Q8K4F0
Gene Alias adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Gene Symbols Cd226
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-254 of human CD226 (NP_0065572).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225825
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.