Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD226 Recombinant Rabbit Monoclonal Antibody (026), FITC
GREENER_CHOICE

Catalog No. PIMA546801
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546801 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546801 Supplier Invitrogen™ Supplier No. MA546801
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD226 (DNAM-1) (Met1-Pro254) and conjugated with FITC under optimum conditions, the unreacted FITC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

DNAM 1 is a receptor expressed by peripheral blood T lymphocytes that is involved in intercellular adhesion, lymphocyte signaling, cytotoxicity and lymphokine secretion mediated by cytotoxic T lymphocytes. DNAM 1 is broadly expressed on T cells, NK cells, platelets, monocytes and a subset of B cells. DNAM-1 is also expressed by a subset of CD3 positive thymocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD226
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 026
Concentration 100 μg/mL
Conjugate FITC
Formulation PBS with 0.5% BSA and 0.09% sodium azide
Gene Cd226
Gene Accession No. Q8K4F0
Gene Alias adhesion glycoprotein; BC051526; Cd226; CD226 antigen; CD226 molecule; DNAM 1; DNAM1; DNAM-1; DNAX accessory molecule 1; DNAX accessory molecule-1; platelet and T cell activation antigen 1; platelet and T-cell activation antigen 1; Pta1; T lineage-specific activation antigen 1 antigen; TLiSA1
Gene Symbols Cd226
Host Species Rabbit
Immunogen Recombinant protein encoding Mouse CD226 (Met1-Pro254).
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225825
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.