Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD229 Monoclonal Antibody (HLy9.25), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA110151
Description
This antibody recognizes an extracellular epitope of CD229 / Ly9, a 100-120 kDa cell surface glycoprotein expressed on T and B cells.
CD229 (Ly9) is a cell surface receptor of the CD150 family, which includes also e.g. CD48 and CD224. Receptors of this family regulate cytokine production and cytotoxicity of lymphocytes and NK cells. High levels of CD229 are found on T and B cells, where its expression increases during their maturation. It is absent on granulocytes, bone marrow-derived dendritic cells, platelets and erythrocytes. CD229 has been also reported on mouse monocytes and NK cells. CD229 interacts homophilically through its N-terminal domain and localizes to the contact site between T cells and antigen presenting B cells during antigen-dependent immune synapse formation.
Specifications
CD229 | |
Monoclonal | |
PE | |
Ly9 | |
AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
4063 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
HLy9.25 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q9HBG7 | |
Ly9 | |
CD299-transfected 300.19 pre-B cell line. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction