Learn More
CD229 Polyclonal Antibody, Invitrogen™
Description
A suggested positive control is 293 cell lysate. PA5-21135 can be used with blocking peptide PEP-1249.
Specifications
Specifications
| Antigen | CD229 |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Ly9 |
| Gene Accession No. | Q01965, Q9HBG7 |
| Gene Alias | AI893573; CD229; CDABP0070; Cell surface molecule Ly-9; hly9; Lgp100; LY9; Ly-9; Lymphocyte antigen 9; mLY9; RP11-312J18.1; Signaling lymphocytic activation molecule 3; SLAM family member 3; SLAMF3; T100; T-lymphocyte surface antigen Ly-9 |
| Gene Symbols | Ly9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.