Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (002), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290617AF647
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD229/SLAMF3/Lymphocyte Antigen 9 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
4063 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
2 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD229/SLAMF3/Lymphocyte Antigen 9 (NP_032560.2; Met1-Arg453). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction