Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (249936), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1898I
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal antibody specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
CD229/SLAMF3/Lymphocyte Antigen 9 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
249936 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
Mouse myeloma cell line NS0-derived recombinant human CD229/SLAMF3, Lys48-Lys454, Accession # Q9HBG7 | |
0.1 mL | |
Immunology | |
4063 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction