Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Rabbit anti-Mouse, Clone: 17, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP29061850UL
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Mouse samples. It is validated for ELISA.Specifications
CD229/SLAMF3/Lymphocyte Antigen 9 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
4063 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA | |
17 | |
ELISA 1:5000-1:10000 | |
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
Recombinant Mouse CD229/SLAMF3/Lymphocyte Antigen 9 protein | |
50 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction