Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (HLy9.25), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP150070JF646
Description
CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD229/SLAMF3/LymphocyteAntigen9 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LY9 | |
CD299-transfected 300.19 pre-B cell line | |
0.1 mL | |
4063 | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
HLy9.25 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 | |
Mouse | |
Protein A purified | |
Primary | |
The mouse monoclonal antibody HLy9.25 (also known as HLy9.1.25) recognizes Lymphocyte Antigen 9, a 100-120 kDa cell surface glycoprotein expressed on T and B cells. | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction