Learn More
CD23/Fc epsilon RII Antibody (MM0164-2C23) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CD23/Fc epsilon RII |
| Applications | CyTOF, Blocking Assay, Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | MM0164-2C23 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Blocking/Neutralizing 50-200 molar excess, CyTOF-ready |
| Formulation | A 0.2 um filtered solution in PBS with No Preservative |
| Gene Accession No. | P06734 |
| Gene Alias | BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.