Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD23/Fc epsilon RII Antibody (OTI2B6), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270701MFV500
Description
CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow CytometrySpecifications
CD23/Fc epsilon RII | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI2B6 | |
Flow Cytometry | |
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor | |
Human recombinant protein fragment corresponding to amino acids 48-321 of human FCER2 (NP_001993) produced in SF9 cell. | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology, Phospho Specific | |
2208 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction