Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23/Fc epsilon RII Rat anti-Mouse, Alexa Fluor 594, Clone: 691632, R&D Systems™

Catalog No. FAB6900T025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6900T025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6900T025 Supplier R&D Systems Supplier No. FAB6900T025
This item is not returnable. View return policy

Rat Monoclonal Antibody

The low affinity receptor for IgE, Fcε RII (designated CD23) is a member of subgroup II of the C-type (Ca2+-dependent) lectin superfamily (1-4). Mouse CD23 is a45-49kDa type II transmembrane glycoprotein that binds mouse, but not human, IgE (1-4). The longest isoform is 331 amino acids (aa) in length and contains a 23 aa cytoplasmic domain, a 26 aa transmembrane segment, and a 282 aa extracellular domain (ECD). The ECD C-type lectin domain binds both protein and carbohydrate, with separate sites for binding IgE, CD21, and β2 and αv integrins (2). Coiled-coil topology within a connecting stalk contributes to oligomerization, which increases IgE affinity (1, 2). Mouse CD23 shares 88% aa sequence identity with rat CD23 and up to 57% aa identity with isoforms of human, equine, porcine and bovine CD23. Isoforms which vary in the cytoplasmic sequence include the “a” isoform, which begins with the sequence MEENEYS and is constitutively expressed by B cells, and the cb” isoform, which begins with MDTHHT and is induced by IL-4 on a variety of inflammatory cells, B cells and epithelia (2, 5-7). The isoforms differentially participate in IgE-mediated endocytosis and phagocytosis (2, 8-10). CD23b and two 309 aa mouse CD23b isoforms, lacking either exon 5 or 6, also display distinct endocytic properties on intestinal epithelia (6, 11). Several soluble forms of CD23 are mainly generated by metalloprotease (especially ADAM10) and cysteine protease activity, although potentially soluble mouse isoforms have also been sequenced (12-14). Both soluble and membrane-bound forms of CD23 include the lectin domain and show bioactivity. CD23 binding to monocyte integrins results in oxidative product generation and proinflammatory cytokine release (15). On human, but not mouse, B cells, sCD23 induces IgE secretion by binding CD21 (2, 9). In both, secreted IgE will bind B cell membrane CD23, rendering it unavailable for cleavage, and thus shutting down IgE production (2).

Specifications

Antigen CD23/Fc epsilon RII
Applications Flow Cytometry
Classification Monoclonal
Clone 691632
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P20693
Gene Alias BLAST-2, CD23, CD23A, CD23CD23 antigen, CLEC4J, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc epsilon RII, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, FCER2, Fcer2a, FceRII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Ly-42, Lymphocyte IgE receptor
Gene Symbols Fcer2a
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD23/Fc epsilon RII Glu50-Pro331 Accession # P20693
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2208
Test Specificity Detects mouse CD23/Fc epsilon RII in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Fc epsilon RIA or rhFc epsilon RIG is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.