Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD23 Mouse anti-Human, BB700, Clone: M-L233, BD Horizon™
Mouse Monoclonal Antibody
$444.00
Specifications
Antigen | CD23 |
---|---|
Clone | M-L233 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow Cytometry.Specifications
CD23 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
M-L233 | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
CD23 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title