Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23 Mouse, anti-Human, R718, Clone: EBVCS-5, BD OptiBuild™

Catalog No. BDB752269 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752269 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752269 Supplier BD Biosciences Supplier No. 752269
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The EBVCS-5 antibody specifically binds to human CD23, the low affinity receptor for human IgE (Fc epsilon RII or FcεRII), which is also known as C-type lectin domain family 4 member J (CLEC4J). CD23 is expressed as a ∼45 kDa type II membrane glycoprotein that is present at low density on most normal B lymphocytes and at higher levels on activated B lymphocytes, Epstein-Barr virus (EBV)-transformed lymphoblasts, chronic lymphocytic leukemia (CLL) cells of B-lymphocyte origin, and tonsillar B lymphocytes. CD23 expression is induced on B cells by interleukin-4 (IL-4) and is lost after isotype switching to IgA, IgG, or IgE. The CD23 antigen is not present on immature bone marrow B lymphocytes or on T lymphocytes. CD23 may also be differentially expressed on monocytes, macrophages, eosinophils, platelets, dendritic cells, and Langerhans cells. CD23 can mediate IgE-dependent cytotoxicity and phagocytosis by macrophages and eosinophils. Soluble CD23 (sCD23) can be released by CD23-positive cells as a result of proteolytic cleavage of membrane CD23. Larger fragments of sCD23 (eg, 25-37 kDa) retain their IgE-binding capacity whereas smaller fragments (ie, ≤ 12 kDa) do not. Soluble CD23 may have immunoregulatory effects on the growth and differentiation of B cells and other cell types.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD23
Applications Flow Cytometry
Classification Monoclonal
Clone EBVCS-5
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.