Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD23, Rabbit Monoclonal Antibody

Catalog No. RM9123R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM9123R7 7 mL Prediluted
RM9123S1 500 μL Concentrated
RM9123S 1 mL Concentrated
RM9123RQ 12 mL Ready-to-Use
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM9123R7 Supplier Epredia™ Supplier No. RM9123R7

Obtain accurate and reproducible results in immunohistochemistry procedures with Epredia™ CD23, Rabbit Monoclonal Antibody.

CD23 is a 45 kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma.

Host Species: Rabbit

Clone: SP23

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant protein encoding human CD23 48-248aa

Molecular Weight: 45 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD23
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP23
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein encoding human CD23 48-248aa
Quantity 7 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.