Learn More
Description
Host Species: Rabbit
Clone: SP23
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant protein encoding human CD23 48-248aa
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD23 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP23 |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encoding human CD23 48-248aa |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Oncogenesis |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.