Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD235a (Glycophorin A) |
---|---|
Clone | HIR2 (GA-R2) |
Concentration | 0.1 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H050T03Y04A
![]() |
Invitrogen™
H050T03Y04A |
100 Tests |
Each for $430.00
|
|
|||||
H050T02Y04A
![]() |
Invitrogen™
H050T02Y04A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD235a (Glycophorin A) Monoclonal antibody specifically detects CD235a (Glycophorin A) in Human samples. It is validated for Flow CytometrySpecifications
CD235a (Glycophorin A) | |
0.1 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P02724 | |
2993 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HIR2 (GA-R2) | |
Flow Cytometry | |
NovaFluor Yellow 660 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
IgG2b κ | |
Affinity chromatography | |
GYPA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title