Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD235a Monoclonal Antibody (JC159)
GREENER_CHOICE

Catalog No. PIMA544034
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544034 Supplier Invitrogen™ Supplier No. MA544034
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody recognizes an epitope between amino acids 27 and 40 of the extracellular portion of CD235a (glycophorin A), a sialoglycoprotein expressed on early erythroblasts, late erythroblasts, erythroblasts, mature erythrocytes and the cells of erythroid cell lines K562 and HEL. The antibody does not react with glycophorin B.

Glycophorin A, also known as CD235a, is a 151 amino acid sialoglycoprotein expressed on the membrane of mature erythrocytes and erythroid progenitor cells, with approximately 500,000 copies per cell. The gene for glycophorin A is located on chromosome 4 and exists in two allelic forms, M and N, which differ by two amino acids: the M group has Ser1 and Gly5, while the N group has Leu1 and Glu5. These allelic variations define the blood group M and N specificities. Glycophorin A serves multiple functions, including providing a mucin-like barrier that minimizes aggregation between red blood cells in circulation, potentially preventing cell fusion. It also acts as a receptor for certain pathogens, including Sandei virus, parvovirus, and Hsa, a Streptococcus adhesin. Recent studies suggest that exposure to toxins can lead to mutations or loss of alleles, resulting in phenotypic changes. There is ongoing research correlating genotype and phenotype with predisposition to diseases such as cancer and heart disease, highlighting the importance of glycophorin A in both health and disease. Glycophorin A is a significant marker for studying erythrocyte-related functions and pathologies, as well as its role in pathogen interactions and disease predisposition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD235a
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JC159
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene GYPA
Gene Accession No. P02724
Gene Alias AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha
Gene Symbols GYPA
Host Species Mouse
Immunogen Membrane preparation from splenic hairy cell leukemia.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2993, 688972
Target Species Human, Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.