Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD235a Monoclonal Antibody (JC159), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA562257

Description
CD235a Monoclonal Antibody for Flow
CD235a (Glycophorins A, GPA) is a single pass membrane sialoglycoprotein expressed in mature erythrocytes and erythroid precursor cells. CD235a is the carrier of blood group M and N specificities. CD235a provides cells with a large mucin-like that may serve as a barrier to cell fusion, minimizing aggregation between red blood cells in the circulation. CD235a has been shown to act as a receptor for Sandei virus, parvovirus, and Hsa, an Streptococcus adhesin.
Specifications
CD235a | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P02724 | |
GYPA | |
Membrane preparation from splenic hairy cell leukemia. | |
100 Tests | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
JC159 | |
PE | |
GYPA | |
AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha | |
Mouse | |
Size-exclusion Chromatography | |
RUO | |
2993, 688972 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction