Learn More
Invitrogen™ CD235a Monoclonal Antibody (YTH89.1), APC

Description
MA5-17697 targets CD235a/Glycophorin A in FACS and WB applications and shows reactivity with Human samples. The MA5-17697 immunogen is rat monoclonal antibodies raised against human CD235a.
Specifications
Specifications
| Antigen | CD235a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | YTH89.1 |
| Concentration | 0.1 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 4-5mg/mL BSA and 0.02% sodium azide |
| Gene | GYPA |
| Gene Accession No. | P02724 |
| Gene Alias | AI853584; CD235; CD235a; erythroid-lineage-specific membrane sialoglycoprotein; glycophorin A; glycophorin A (MN blood group); glycophorin A (MNS blood group); glycophorin A, GPA; glycophorin Erik; glycophorin MiI; glycophorin MiV; glycophorin SAT; glycophorin Sta type C; glycophorin-A; GPA; GPErik; GPSAT; Gypa; HGNC:4702; HGpMiV; HGpMiX; HGpMiXI; HGpSta(C); HGpStaC; Mi.V glycoprotein; Mi.V glycoprotein (24 AA); MN; MN sialoglycoprotein; MNS; PAS-2; recombinant glycophorin A-B Miltenberger-DR; sialoglycoprotein alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.