Learn More
CD235a Mouse anti-Human, APC, Clone: GA-R2 (HIR2), BD
Mouse Monoclonal Antibody
$188.00 - $390.00
Specifications
Antigen | CD235a |
---|---|
Clone | GA-R2 (HIR2) |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561775
|
BD Biosciences
561775 |
25 μg |
Each for $188.00
|
|
|||||
BDB551336
|
BD Biosciences
551336 |
0.1 mg |
Each for $390.00
|
|
|||||
Description
The GA-R2 (also known as HIR2) monoclonal antibody specifically binds to CD235a, otherwise known as Glycophorin A (GYPA, GPA), PAS-2, MN sialoglycoprotein (MNS, MN), and Sialoglycoprotein A/alpha. CD235a is a type I transmembrane sialoglycoprotein that is expressed on human erythrocytes, erythroid precursor cells and certain leukemic cell types. This antibody is useful for the identification and characterization of erythrocytes, certain myeloid leukemic cell types, and studies of erythroid cell development and infectious diseases with erythrocyte involvement. Mature, non-nucleated red blood cells characteristically express very high levels of glycophorin A.
Specifications
CD235a | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Human | |
IgG2b κ | |
Affinity Purified | |
Glycophorin-A; GYPA; GPA; PAS-2; MN sialoglycoprotein; Sialoglycoprotein A |
GA-R2 (HIR2) | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.