Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD235a Mouse anti-Human, BUV395, Clone:  GA-R2 (HIR2), BD Horizon™

Catalog No. BDB565973 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565973 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565973 Supplier BD Biosciences Supplier No. 565973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD235a Monoclonal antibody specifically detects CD235a in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD235a
Applications Flow Cytometry
Classification Monoclonal
Clone  GA-R2 (HIR2)
Conjugate Brilliant Ultraviolet 395
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Glycophorin-A; GYPA; GPA; PAS-2; MN sialoglycoprotein; Sialoglycoprotein A
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions
Form Purified
Isotype IgG2b κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.