Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Antibody (4F4E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$572.50
Specifications
Antigen | CD24 |
---|---|
Clone | 4F4E10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
Description
CD24 Monoclonal specifically detects CD24 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
CD24 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
RUO | |
Human | |
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
CD24 | |
IgG1 | |
Affinity Purified |
4F4E10 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Mouse | |
Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
100133941 | |
Purified recombinant fragment of human CD24 (AA: 15-80) expressed in E. Coli. | |
Primary | |
8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title