Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Antibody (910019), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB8547APCCY7
Description
CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD24 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 910019 | |
| Flow Cytometry | |
| CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
| Peptide sequence spanning a portion of the extracellular region, Accession # P24807 | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Stem Cell Markers | |
| 100133941 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction