Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Antibody (ML5), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5247I
This item is not returnable.
View return policy
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow CytometrySpecifications
CD24 | |
Monoclonal | |
Janelia Fluor 549 | |
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
Human CD24 | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Stem Cell Markers | |
100133941 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
ML5 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction