Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Antibody (SN3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064861H
Description
CD24 Monoclonal specifically detects CD24 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD24 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CD24 | |
Glycoproteins purified from human NALM-1 cell line | |
0.1 mL | |
B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
100133941 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
SN3 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes the human CD24 cell surface antigen, a heavily glycosylated molecule of 35-45kD. CD24 is expressed by granulocytes and B lymphocytes, and by some activated T cells and T cell malignancies. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction