Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Antibody (SR1522), Novus Biologicals™

Rabbit Monoclonal Antibody
$413.50
Specifications
Antigen | CD24 |
---|---|
Clone | SR1522 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
CD24 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
100133941 | |
Affinity purified |
SR1522 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
B Cell Development and Differentiation Markers, Cancer, Neuronal Cell Markers, Stem Cell Markers | |
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title