Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ CD24 (GPI-Linked Surface Mucin) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1279P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
Concentration:
1mg/mL
200μg/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS1279P0 100 μL 200μg/mL
MS1279P1 500 μL 200μg/mL
MS1279P1ABX 100 μL 1mg/mL
MS1279PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1279P0 Supplier Epredia™ Supplier No. MS1279P0

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures.

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. It is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin's lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation.

Host Species: Mouse

Clone: SN3b

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line

Molecular Weight: 30-70 kDa

Positive Control: Breast CA, Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry

Note:

USA: IVD; Int'l: IVD

Specifications

Antigen CD24 (GPI-Linked Surface Mucin) Ab-2
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN3b
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Cell membrane antigen preparation isolated from NALM-1 human pre-B leukemia cell line
Quantity 100 μL
Regulatory Status RUO
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.