Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Monoclonal Antibody (eBioSN3 (SN3 A5-2H10)), NovaFluor™ Red 725, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD24 |
---|---|
Clone | eBioSN3 (SN3 A5-2H10) |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H064T03R05A
![]() |
Invitrogen™
H064T03R05A |
100 Tests |
Each for $430.00
|
|
|||||
H064T02R05A
![]() |
Invitrogen™
H064T02R05A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD24 Monoclonal antibody specifically detects CD24 in Human samples. It is validated for Flow CytometrySpecifications
CD24 | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P25063 | |
100133941 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBioSN3 (SN3 A5-2H10) | |
Flow Cytometry | |
NovaFluor Red 725 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen | |
IgG1 κ | |
Affinity chromatography | |
Cd24 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title