Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Monoclonal Antibody (M1/69), Functional Grade, In Vivo GOLD™, Leinco Technologies™
SDP

Catalog No. 502544237
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4237 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4237 Supplier Leinco Technologies Inc Supplier No. C333100MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specificity: Anti-CD24 antibody (clone M1/69) activity is directed against mouse CD24, also known as Heat Stable Antigen (HSA) or Ly-52. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -70C. Avoid Repeated Freeze Thaw Cycles. Antibody has been tested in Indirect ELISA.

CD24 Monoclonal antibody specifically detects CD24 in Mouse samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Inhibition Assays, Western Blot

Specifications

Antigen CD24
Applications ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Inhibition Assays, Western Blot
Classification Monoclonal
Clone M1/69
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene Cd24a
Gene Accession No. P24807
Gene Alias CD24; CD24 antigen; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 molecule; CD24A; CD24a antigen; cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; heat-stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; Nectadrin; nectadrin heat stable antigen; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen
Host Species Rat
Immunogen C57BL/10 mouse splenic T cells
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12484
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.