Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Mouse anti-Human, Clone: ML5, Abnova™

Catalog No. 89933762 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-762 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-762 Supplier Abnova Corporation Supplier No. MAB12201

Mouse monoclonal antibody raised against full length recombinant CD24.

Specifications

Antigen CD24
Applications Immunofluorescence
Classification Monoclonal
Clone ML5
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Gene Alias CD24A, FLJ22950, FLJ43543, MGC75043
Gene Symbols CD24
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CD24.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100133941
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.