Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Mouse anti-Human, FITC, Clone: ML5, BD

Catalog No. BDB555427 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555427 100 Tests
BDB560992 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555427 Supplier BD Biosciences Supplier No. 555427
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ML5 monoclonal antibody specifically binds to CD24. CD24 is a 35-45 kDa two-chain glycoprotein expressed on the surface of B cells, granulocytes and most B-cell lines. CD24 may play a role in the regulation of B-cell proliferation and maturation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD24
Applications Flow Cytometry
Classification Monoclonal
Clone ML5
Conjugate FITC
Description Heat Stable Antigen Homologue (HSA); Ba-1; CD24A
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.