Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Mouse, anti-Human, R718, Clone: ML5, BD OptiBuild™

Catalog No. BDB751874 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751874 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751874 Supplier BD Biosciences Supplier No. 751874
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ML5 monoclonal antibody specifically binds to CD24 which is also known as CD24A, signal transducer CD24, small cell lung carcinoma cluster 4 antigen, or BA-1. CD24 is a 35-70 kDa glycophosphatidylinositol (GPI)-linked glycoprotein whose glycosylation pattern is highly variable and cell-type dependent. CD24 is expressed on neutrophils, eosinophils, dendritic cells, neural cells, epithelial cells, muscle cells, and many types of cancer cells. CD24 functions as an adhesion receptor. Several different ligands have been identified for CD24 including CD62P (P-selectin) which is expressed on activated platelets and activated endothelium. CD24 is variably expressed on all B lineage cells, except plasma cells, and can play a role in regulating the activation, proliferation or differentiation of these cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD24
Applications Flow Cytometry
Classification Monoclonal
Clone ML5
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.