Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD243 Mouse, anti-Human, R718, Clone: UIC2, BD OptiBuild™

Catalog No. BDB751945 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751945 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751945 Supplier BD Biosciences Supplier No. 751945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The UIC2 monoclonal antibody specifically binds to an extracellular epitope of CD243, which is also known as ATP-binding cassette subfamily B member 1 (ABCB1), Multidrug resistance protein 1 (MDR1), or P-glycoprotein 1. CD243 is a transmembrane glycoprotein that spans the membrane 12 times. CD243 acts as an ATP-dependent efflux pump for a large variety of lipophilic molecules and drugs. This efflux activity has been suggested to lead to resistance to the drugs used in chemotherapy. CD243 is expressed by epithelial and endothelial cells, and at low levels by T cells, B cells, NK cells, and hematopoietic stem cells. It may be expressed at high levels by multidrug resistant (MDR) tumor cells. The UIC2 antibody reportedly inhibited the efflux of CD243 substrates from MDR cells and increased the cytotoxicity of certain CD243-transported drugs. The UIC2 antibody reportedly does not crossreact with mouse CD243.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD243
Applications Flow Cytometry
Classification Monoclonal
Clone UIC2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.