Learn More
Invitrogen™ CD244.1 Monoclonal Antibody (C9.1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
Supplier: Invitrogen™ 46244082

Description
This gene encodes a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via this receptor is thought to modulate NK-cell cytolytic activity. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
This gene encodes a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via this receptor is thought to modulate NK-cell cytolytic activity. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
CD244.1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q07763 | |
Cd244 | |
Affinity chromatography | |
RUO | |
18106 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
C9.1 | |
PerCP-eFluor 710 | |
Cd244 | |
2B4; C9.1; CD244; CD244 long isoform; CD244 natural killer cell receptor 2B4; CD244 short isoform; F730046O15Rik; Ly90; NAIL; natural killer cell receptor; Natural killer cell receptor 2B4; NK cell receptor 2B4; NK cell type I receptor protein 2B4; NKR2B4; Nmrk; non MHC restricted killing associated; non-MHC restricted killing associated; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.