Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD244 (2B4) Monoclonal Antibody (M085)
GREENER_CHOICE

Catalog No. PIMA547660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547660 Supplier Invitrogen™ Supplier No. MA547660
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone M085 mouse monoclonal antibody detects a 110 kDa* protein on SDS-PAGE Native immunoblots of human THP1 monocytes. The antibody detects only the native CD244 protein, and the aldehyde-fixed CD244 protein. The antibody works for native western blot, immunoprecipitation, immunocytochemistry, and ELISA capture.

CD244 (Natural killer (NK) cell receptor 2B4/SLAMF4) is an Ig superfamily signaling lymphocyte activation molecule (SLAM) receptor. Like all SLAM family receptors, it has an extracellular segment with two immunoglobulin (Ig)-like domains, and a cytoplasmic domain containing four immunoreceptor tyrosine-based switch motifs. CD244 does not act as a selfligand similar to other SLAM family receptors. It binds CD48, a transmembrane receptor ubiquitously expressed on hematopoietic cells. CD244 activity is controlled by the presence or absence of small cytoplasmic adapter proteins, SH2D1A/SAP and/or SH2D1B/EAT-2. Downstream signaling involves predominantly VAV1, and, to a lesser degree, INPP5D/SHIP1 and CBL. Activation of CD244 stimulates NK cell cytotoxicity, production of IFN-gamma and granule exocytosis. CD244 is involved in the regulation of CD8+ T-cell proliferation, and inhibits inflammatory responses in dendritic cells (DCs). In cancers, CD244 shows increased expression in intratumoral DCs and myeloid suppressor cells, and anti-CD244 therapies may increase infiltrating T-cells and impair tumor growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD244 (2B4)
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M085
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene CD244
Gene Accession No. Q9BZW8
Gene Alias NKR2B4; SLAMF4
Gene Symbols CD244
Host Species Mouse
Immunogen Clone (M085) was generated from a recombinant protein that included the extracellular region of human CD244 protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51744
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.