Learn More
Invitrogen™ CD247 (CD3 zeta) Monoclonal Antibody (6B10.2), PerCP-eFluor™ 710, eBioscience™

Description
Description: This 6B10.2 monoclonal antibody reacts with human CD247, which is also known as the invariant zeta chain. The CD3 complex is composed of three monomeric chains (gamma, delta, and epsilon) and the invariant zeta chain, which exists as a homodimer or as a heterodimer with the epsilon subunit. Together with the T cell antigen receptor (TCR), the CD3 gamma, delta, epsilon, and zeta chains comprise the TCR/CD3 complex. CD3 zeta is expressed on immature and mature thymocytes, as well as T lymphocytes and NK cells. Signal transduction during T cell activation is mediated by tyrosine phosphorylation of cytoplasmic immunoreceptor tyrosine-based activation motifs (ITAMs) present on the zeta chain. The 6B10.2 monoclonal antibody recognizes a transmembrane epitope of the CD3 zeta chain. Therefore, flow cytometric analysis with this antibody requires intracellular staining. In addition, this antibody has been reported to recognize the zeta chain as a homodimer but not as a monomer. Applications Reported: This 6B10.2 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 6B10.2 antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (Product # 88-8824-00) and protocol. Please refer to BestProtocols®: Protocol A: Two step protocol for (cytoplasmic) intracell...
Specifications
Specifications
| Antigen | CD247 (CD3 zeta) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 6B10.2 |
| Concentration | 5 μL/Test |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD247 |
| Gene Accession No. | P20963 |
| Gene Alias | 4930549J05Rik; A430104F18Rik; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; Cd3; CD3 antigen, zeta polypeptide; CD3 zeta; CD3 zeta chain; CD3e antigen; CD3-epsilon; Cd3-eta; CD3H; CD3Q; Cd3z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; Cd3-zeta; CD3zeta chain; CD3-zeta/eta; FLJ18683; IMD25; T3E; T3z; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 eta chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface glycoprotein CD3 zeta chain; TCR zeta chain; TCRE; Tcrk; TCRZ; TCRzeta |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.