Learn More
CD247/CD3Z Recombinant Rabbit Monoclonal Antibody (BL-336-1B2), Bethyl Laboratories
Rabbit Recombit Monoclonal Antibody
$157.08 - $595.98
Specifications
Antigen | CD247/CD3Z |
---|---|
Clone | BL-336-1B2 |
Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
Classification | Recombinant Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1288
![]() |
Bethyl Laboratories, Inc
A700017T |
10 μL |
Each for $157.08
|
|
|||||
50-157-1287
![]() |
Bethyl Laboratories, Inc
A700017 |
100 μL |
Each for $595.98
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.Specifications
CD247/CD3Z | |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P20963 | |
919 | |
Between 114 and 164 | |
Primary | |
4° C | |
CD247 |
BL-336-1B2 | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2 | |
T3Z, CD3H, CD3Q, TCRZ, CD3-ZETA | |
CD247 | |
IgG | |
Purified | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.