Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD248 Monoclonal Antibody (OTI1H6)
GREENER_CHOICE

Catalog No. PIMA526838
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526838 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526838 Supplier Invitrogen™ Supplier No. MA526838
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD248 Monoclonal Antibody for Western Blot, IHC (P)

CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD248
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD248
Gene Accession No. Q9HCU0
Gene Alias 2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1
Gene Symbols CD248
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 20-300 of CD248 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57124
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.