Learn More
Invitrogen™ CD248 Polyclonal Antibody

Description
A suggested positive control is human colon tissue lysate. PA5-20437 can be used with blocking peptide PEP-0554. The PA5-20437 immunogen is located within amino acids 40-90 of TEM-1. Predicted molecular ~ 81kD. In Western blot applications, this antibody has been observed to detect a band at: 90 kD (post-modifications: 27 O-linked glycosylations). Predicted species reactivity based on immunogen sequence: Mouse (100%); Rat: (93%).
Specifications
Specifications
| Antigen | CD248 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CD248 |
| Gene Accession No. | Q9HCU0 |
| Gene Alias | 2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1 |
| Gene Symbols | CD248 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.