Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD249 (Ly-51, 6C3/BP-1 antigen) Mouse anti-Mouse, BUV615, Clone: BP-1, BD OptiBuild™

Catalog No. BDB752355 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752355 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752355 Supplier BD Biosciences Supplier No. 752355
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD249 (Ly-51, 6C3/BP-1 antigen) Monoclonal antibody specifically detects CD249 (Ly-51, 6C3/BP-1 antigen) in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD249 (Ly-51, 6C3/BP-1 antigen)
Applications Flow Cytometry
Classification Monoclonal
Clone BP-1
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms CD249 (Ly-51) BUV615 BP-1 50 μg
Host Species Mouse
Immunogen Abelson murine leukemia virus-transformed BALB/c mouse pre-B cell line 18.81
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD249 (Ly-51, 6C3/BP-1 antigen)
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.