Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD249 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588487 Supplier Invitrogen™ Supplier No. PA588487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFTVIRYISY NSYGKNMAWN WIQLNWDYLV NRYTLNNRNL GRIVTIAEPF NTELQLWQME SFFAKYPQAG AGEKPREQVL ETVKNNIEWL KQHRNTIREW F; Positive Samples: 293T, LO2, HT-29, Mouse liver; Cellular Location: Membrane, Single-pass type II membrane protein.

CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD249
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Enpep
Gene Accession No. P16406, Q07075
Gene Alias 6030431M22Rik; Aminopeptidase A; aminopeptidase-A; APA; AP-A; BP1; Bp-1/6C3; BP-1/6C3 antigen; CD249; differentiation antigen gp160; EAP; ENPEP; Glutamyl aminopeptidase; glutamyl aminopeptidase (aminopeptidase A); gp160; Ly51; Ly-51
Gene Symbols Enpep
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13809, 2028
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.