Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25 (IL-2 Receptor α), Mouse Anti-Human, RB613, 2A3, BD Horizon™
Mouse Monoclonal Antibody
$164.00 - $406.00
Specifications
Antigen | CD25 (IL-2 Receptor α) |
---|---|
Clone | 2A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB613 |
Description
CD25 (IL-2 Receptor α) Monoclonal antibody specifically detects CD25 (IL-2 Receptor α) in Human (QC Testing) samples. It is validated for Flow CytometrySpecifications
CD25 (IL-2 Receptor α) | |
Flow Cytometry | |
RB613 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3559 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2A3 | |
Monoclonal | |
Mouse | |
Human | |
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 | |
Human Phytohemagglutinin-activated T Cells | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title