Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2Rα Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS203P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS203P1 500 μL 200μg/mL
MS203P0 100 μL 200μg/mL
MS203P1ABX 100 μL 1mg/mL
MS203P 1 mL 200μg/mL
MS203PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS203P1 Supplier Epredia Supplier No. MS203P1
Add to Cart
Add to Cart

Obtain accurate, reproducible results in immunofluorescence and flow cytometry experiments with Epredia™ CD25/IL-2Rα Ab-1, Mouse Monoclonal Antibody.

IL-2R (T-cell growth factor receptor) consists of a chain of 55kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages.

Host Species: Mouse

Clone: IL2R.1

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Recombinant human interleukin-2 receptor

Molecular Weight: 55 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD25/IL-2Ralpha Ab-1
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone IL2R.1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human interleukin-2 receptor
Quantity 500 μL
Regulatory Status RUO
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.