Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD25/IL-2Rα Ab-4, Mouse Monoclonal Antibody

Catalog No. MS1088S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1088S1 500 μL Concentrated
MS1088S 1 mL Concentrated
MS1088R1 1 mL Prediluted
MS1088R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1088S1 Supplier Epredia™ Supplier No. MS1088S1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD25/IL-2Rα Ab-4, Mouse Monoclonal Antibody.

IL-2R (T-cell growth factor receptor) consists of a chain of 55 kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages. CD25 molecule alone appears to function as a low affinity receptor and associates wtih CD122 (IL-2R Β chain, p75) and CD132 (common γ chain) to form the high affinity IL-2 receptor complex

Host Species: Mouse

Clone: 4C9

Isotype: IgG2b

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein corresponding to the external domain of the Interleukin-2 Receptor molecule

Molecular Weight: 55 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD25/IL-2Ralpha Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C9
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to the external domain of the Interleukin-2 Receptor molecule
Purification Method Tonsil or lymph node
Quantity 500 μL
Regulatory Status RUO
Research Discipline B-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.