Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Antibody (051), DyLight 488

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290518G
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
Monoclonal | |
DyLight 488 | |
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD25/IL-2R alpha (NP_032393.3; Met1-Lys236). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
3559 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
51 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction