Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Antibody (280402) - Azide Free, Novus Biologicals™

Rat Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | CD25/IL-2R alpha |
---|---|
Clone | 280402 |
Concentration | 0.36 mg/mL |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB24382SP
![]() |
Novus Biologicals
MAB24382SP |
25 μg |
Each for $181.90
|
|
|||||
MAB24382
![]() |
Novus Biologicals
MAB24382 |
100 μg |
Each for $487.50
|
|
|||||
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
0.36 mg/mL | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
PBS with No Preservative | |
3559 | |
Partial recombinant mouse CD25/IL2Ra protein (amino acids 22-236) [UniProt P01590]. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
280402 | |
Flow Cytometry | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
IL2RA | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title