Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Antibody (280406), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2438H
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 280406 | |
| Flow Cytometry, CyTOF-ready | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD25/IL-2 R alpha , Glu22-Lys236, Accession # P01590 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
| 3559 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction