Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Antibody (Daclizumab), Alexa Fluor™ 594, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275901AF594
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
CD25/IL-2R alpha | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
IL2RA | |
Cultured T-cells derived from a patient with Mycosis fungoides. | |
Primary | |
Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
Daclizumab | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
Human | |
0.1 mL | |
3559 | |
Human, Cynomolgus Monkey, Rhesus Macaque | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction