Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Antibody (Daclizumab), DyLight 550, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275901R
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IL2RA | |
| Cultured T-cells derived from a patient with Mycosis fungoides. | |
| 0.1 mL | |
| Primary | |
| Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| Daclizumab | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Human | |
| Protein A purified | |
| Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
| 3559.0 | |
| Human, Cynomolgus Monkey, Rhesus Macaque | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction