Learn More
CD25 (IL-2R alpha) Monoclonal Antibody (7G7B6), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc C9305100MG


Description
Specificity: Clone 7G7B6 reacts with human IL-2Ralpha. Endotoxin Level: <0.5 EU/mg as determined by the LAL method. Purity: ≥98% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
CD25 (IL-2R alpha) Monoclonal antibody specifically detects CD25 (IL-2R alpha) in Human samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
CD25 (IL-2R alpha) | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P01589 | |
Mouse | |
Protein A/G | |
RUO | |
3559 | |
4°C short term, -80°C long term | |
Liquid |
ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry, Immunoprecipitation, Western Blot | |
7G7B6 | |
Functional Grade | |
IL2RA | |
ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
Recombinant human peripheral blood mononuclear cells (PBMCs) | |
100 mg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.